Learn More
Invitrogen™ CHAMP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588213
Description
Immunogen sequence: LPKSALFSES QKAVELGDEL QIDAIDDQKC DILVQEELLA SPKKLLEDTL FPSSKKLKKD NQESSDAELS SSEYIKTDLD AMDIKGQESS SDQEQVDVES IDFSKENKMD MTSPEQSRNV LQFTEEKEAF ISEEEIAKYM KRGKGKYYCK ICCCRAMKKG AVLHHLVNKH NVHSPYKCTI CGKAFLLESL LKNHVAAHGQ SLLKCPRCNF ESNFPRGFKK HLTHCQSRHN EEANKKLMEA LEPPLEEQQI; Positive Samples: U-87MG, A-431, Jurkat ; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, cytoskeleton, kinetochore, spindle.
Required for proper alignment of chromosomes at metaphase and their accurate segregation during mitosis. Involved in the maintenance of spindle microtubules attachment to the kinetochore during sister chromatid biorientation. May recruit CENPE and CENPF to the kinetochore.
Specifications
| CHAMP1 | |
| Polyclonal | |
| Unconjugated | |
| CHAMP1 | |
| AA675043; AI116001; C13orf8; CAMP; CHAMP; Champ1; chromosome alignment maintaining phosphoprotein 1; chromosome alignment-maintaining phosphoprotein 1; D8Ertd457e; D8Ertd569e; Kiaa1802; mKIAA1802; MRD40; RGD1311283; Zfp828; zinc finger protein 828; ZNF828 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283489 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96JM3 | |
| CHAMP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 563-812 of human CHAMP1 (NP_1158121). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.