Learn More
LI-COR Biosciences™ Chameleon Duo Pre-stained Protein Ladder
Description
Capture the sharp, vibrant colors of Chameleon Protein Ladders. In gels, the Ladder can be used to visualize the progress of the protein separation during electrophoresis and to estimate the molecular weight of unknown proteins based on their relative mobility. In blotting applications, the Ladder can be used to monitor protein transfer and as a reference to estimate molecular weight of proteins of interest.
- Provides multi-colored, pre-stained bands for detection via visual inspection or in near-infrared fluorescent bands (700nm or 800nm channels)
- Recombinant proteins in convenient and consistent sizes (8 to 260 kDa) for broad-range molecular weight estimation
- Use for easy visualization of gel migration and protein size, and to orient the gel or membrane quickly
- Ladders are optimized for use with Bis-Tris and Tris-Glycine gels
Specifications
Specifications
| Color | Blue, Red |
| Form | Liquid |
| Formulation | Glycerol; (R,R)-(+)-1,4-dimercaptobutane 2,3 diol; Sodium doceyl sulfate in Water |
| For Use With (Application) | Western blot or gels: visualization of gel migration and protein size, and to orient the gel or membrane |
| No. of Bands | 11 |
| Quantity | 500 μL |
| Range | 8 to 260 kDa |
| Storage Requirements | Frozen (-15 to -25°C), in small aliquots |
| Product Type | Protein Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.