missing translation for 'onlineSavingsMsg'
Learn More

Chalcone alpha,beta-epoxide, 98%

Product Code. 11386686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386686 5 g 5g
11396686 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386686 Supplier Thermo Scientific Alfa Aesar Supplier No. L03874.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5411-12-1 | C15H12O2 | 224.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5411-12-1
MDL Number MFCD00022336

Specifications

Melting Point 88°C to 90°C
Molecular Formula C15H12O2
Quantity 5 g
Beilstein 164553
Synonym 2-benzoyl-3-phenyloxirane, chalcone oxide, 1,3-diphenyl-2,3-epoxy-1-propanone, chalcone epoxide, trans-chalcone oxide, phenyl 3-phenyloxiran-2-yl methanone, ccris 2214, trans-phenyl 3-phenyloxiranyl methanone, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone
InChI Key UQGMJZQVDNZRKT-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3
IUPAC Name phenyl-(3-phenyloxiran-2-yl)methanone
Molecular Weight (g/mol) 224.26
PubChem CID 92219
Formula Weight 224.26
Percent Purity 98%
Chemical Name or Material Chalcone alpha,beta-epoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.