missing translation for 'onlineSavingsMsg'
Learn More

Chalcogran, 96%, mixture of isomers

Product Code. 15761769 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15761769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15761769 Supplier Thermo Scientific Acros Supplier No. 461410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38401-84-2 | C9H16O2 | 156.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38401-84-2
Quantity 1 g
Packaging Glass Bottle
Assay Percent Range 95% min. (GC)
Molecular Formula C9H16O2
Synonym 2-ethyl-1,6-dioxaspiro 4.4 nonane, 2-ethyl-1,6-dioxaspiro 4.4-nonane, 2-ethyl-1,6-dioxaspiro 4.4 nonan, ethyl-1,6-dioxaspiro 4.4-nonane, 8-ethyl-4,9-dioxaspiro 4.4 nonane, 2-ethyl-1,6-dioxaspiro 4,4 nonane, 1,6-dioxaspiro 4.4 nonane, 2-ethyl, 2-ethyl-1,6-dioxaspiro 4.4 nonane #, 2-ethyl-1,6-dioxaspiro 4,4 nonane gc
Boiling Point 67.0°C to 69.0°C
InChI Key DCWKALWZHORJAO-UHFFFAOYSA-N
SMILES CCC1CCC2(O1)CCCO2
IUPAC Name 2-ethyl-1,6-dioxaspiro[4.4]nonane
Molecular Weight (g/mol) 156.22
PubChem CID 581383
Formula Weight 156.22
Percent Purity 96%
Chemical Name or Material Chalcogran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.