missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAF1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17037-1-AP-20UL
This item is not returnable.
View return policy
Description
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro.Specifications
| CHAF1A | |
| Polyclonal | |
| Unconjugated | |
| CHAF1A | |
| CAF, CAF 1, CAF 1 subunit A, CAF I 150 kDa subunit, CAF I p150, CAF1, CAF1B, CAF1P150, CHAF1A, hp150, P150 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10036, 27221 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13111, Q9QWF0 | |
| CHAF1A | |
| CHAF1A Fusion Protein Ag10536 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction