missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Chaetocin
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Description
- Chaetocin is a fungal mycotoxin, originally produced by Chaetomium species, that inhibits the lysine-specific histone methyltransferases (HMTs) SU(VAR)3-9 (IC50 = 0.8 μM)
- HMTs are responsible for methylation of histones, thereby affecting heterochromatinization
- Chaetocin is also known to inhibit G9a and DIM5 (IC50 = 2.5 μM and 3 μM respectively; Cherblanc et al.; Greiner et al.) Chaetocin also acts as a competitive substrate and inhibitor of the central oxidative stress remediation enzyme thioredoxin reductase-1 (TrxR1; Km = 4.6 μM) more potently than glutathione reductase or thioredoxin (Tibodeau et al.)
- Induces cellular oxidative stress, selectively killing cancer cells and rapidly-proliferating primary cells (Isham et al.; Spannhoff et al.)
- Attenuates the growth of glioma xenografts accompanied by an increase in reactive oxygen species (ROS) production (Dixit et al.)
Specifications
Specifications
| CAS | 28097-03-2 |
| Synonym | Chaetocin A |
| Quantity | 500 μg |
| Molecular Formula | C30H28N6O6S4 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction