missing translation for 'onlineSavingsMsg'
Learn More

CHADL Antibody, PerCP, R&D Systems™

Product Code. 18600071 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18600071 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18600071 Supplier Novus Biologicals Supplier No. NBP281906PCP
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

CHADL Polyclonal specifically detects CHADL in Human/Mouse/Rat samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHADL
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias chondroadherin-like, chondroadherin-like protein
Host Species Rabbit
Immunogen CHADL antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human CHADL. The immunogen is located within the last 50 amino acids of CHADL. Amino Acid Squence: ARKAKRTPASRPSARRTPI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150356
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.