missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553761
This item is not returnable.
View return policy
Description
Immunogen sequence: LRWLYLSENA LSSLQPGALD DVENLAKFHV DRNQLSSYPS AALSKLRVVE ELKLSHNPLK SIPDNAFQSF GRYLETLWLD NTNLEKFSDG AFLGVTT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
Specifications
| CHAD | |
| Polyclonal | |
| Unconjugated | |
| CHAD | |
| Cartilage leucine-rich protein; Chad; Chondroadherin; chondroadherin proteoglycan; SLRR4A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15335 | |
| CHAD | |
| Recombinant protein corresponding to Human CHAD. Recombinant protein control fragment (Product #RP-91869). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction