missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561914
This item is not returnable.
View return policy
Description
Immunogen sequence: LGPAPLEDIA EQIFNAAGPS GRNTEYLFEL ANSIRNLVPE EADEHLFALE KLVKERLEGK QNLN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
CHAC2 catalyzes the cleavage glutathione into 5-oxoproline and a Cys-Gly dipeptide and acts specifically on glutathione, but not on other gamma-glutamyl peptides.
Specifications
| CHAC2 | |
| Polyclonal | |
| Unconjugated | |
| Chac2 | |
| cation transport regulator-like protein 2; ChaC cation transport regulator homolog 2; ChaC, cation transport regulator 2; ChaC, cation transport regulator homolog 2; ChaC, cation transport regulator homolog 2 (E. coli); ChaC, cation transport regulator-like 2; Chac2; CHAC2 (HGNC:32363); Gamma-GCG 2; gamma-GCT acting on glutathione homolog 2; Glutathione-specific gamma-glutamylcyclotransferase 2; Putative glutathione-specific gamma-glutamylcyclotransferase 2; RGD1309120 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 494143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUX2 | |
| Chac2 | |
| Recombinant protein corresponding to Human CHAC2. Recombinant protein control fragment (Product #RP-99656). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction