missing translation for 'onlineSavingsMsg'
Learn More

ChABC Antibody (6A12), PE, Novus Biologicals™

Product Code. 18411961 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411961 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411961 Supplier Novus Biologicals Supplier No. NBP196142PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ChABC Monoclonal antibody specifically detects ChABC in Nonspecies specific samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChABC
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6A12
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias Chondroitinase ABC
Host Species Mouse
Immunogen Chondroitinase ABC
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.