missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChABC Antibody (1E10), mFluor Violet 610 SE, Novus Biologicals™
Description
ChABC Monoclonal antibody specifically detects ChABC in Rat, Nonspecies specific samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | ChABC |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1E10 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chondroitinase ABC |
| Host Species | Mouse |
| Immunogen | Chondroitinase ABC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?