missing translation for 'onlineSavingsMsg'
Learn More

CH7057288, MedChemExpress

Product Code. 16470467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470467 50 mg 50mg
16432956 25 mg 25mg
16426817 5 mg 5mg
16425086 100 mg 100mg
16404118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470467 Supplier Medchem Express Supplier No. HY107362

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CH7057288 is a potent and selective TRK inhibitor.

CH7057288 is a potent and selective TRK inhibitor.

Specifications

Chemical Name or Material CH7057288
CAS 2095616-82-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H31N3O5S
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (59.68 mM)
SMILES O=C1C2=C(OC3=CC(C#CC4=CC(C(NC(C)(C)C)=O)=CC(C)=N4)=CC=C23)C(C)(C)C5=CC(NS(C)(=O)=O)=CC=C15
Molecular Weight (g/mol) 569.67
Formula Weight 569.67
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.