missing translation for 'onlineSavingsMsg'
Learn More

CH5424802 (Alectinib) Hydrochloride, TRC

Product Code. 30134684
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134684 50 mg 50mg
30004094 5 mg 5mg
30005347 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134684 Supplier Toronto Research Chemicals Supplier No. TRCC18336550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256589-74-8
Molecular Formula C30 H34 N4 O2 . Cl H
Molecular Weight (g/mol) 519.08
Synonym 5H-Benzo[b]carbazole-3-carbonitrile, 9-ethyl-6,11-dihydro-6,6-dimethyl-8-[4-(4-morpholinyl)-1-piperidinyl]-11-oxo-, hydrochloride (1:1), Alectinib hydrochloride
IUPAC Name 9-ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[b]carbazole-3-carbonitrile;hydrochloride
SMILES Cl.CCc1cc2C(=O)c3c([nH]c4cc(ccc34)C#N)C(C)(C)c2cc1N5CCC(CC5)N6CCOCC6

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C30H34N4O2.ClH/c1-4-20-16-23-24(17-26(20)34-9-7-21(8-10-34)33-11-13-36-14-12-33)30(2,3)29-27(28(23)35)22-6-5-19(18-31)15-25(22)32-29;/h5-6,15-17,21,32H,4,7-14H2,1-3H3;1H
Formula Weight 518.2449
Chemical Name or Material Alectinib hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.