missing translation for 'onlineSavingsMsg'
Learn More

CH5132799, MedChemExpress

Product Code. 16499127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499127 5 mg 5mg
16476548 10 mg 10mg
16466776 100 mg 100mg
16402647 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499127 Supplier Medchem Express Supplier No. HY15466
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CH5132799 is a selective class I PI3K inhibitor. CH5132799 inhibits class I PI3Ks, particularly PI3Kα, with an IC50 of 14 nM.

CH5132799 is a selective class I PI3K inhibitor. CH5132799 inhibits class I PI3Ks, particularly PI3Kα, with an IC50 of 14 nM.

Specifications

Chemical Name or Material CH5132799
CAS 1007207-67-1
Color Gray
Purity Grade Notes Research
Molecular Formula C15H19N7O3S
Quantity 5 mg
Solubility Information DMSO : 4.55 mg/mL (12.06 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=C3C(N(S(=O)(C)=O)CC3)=NC(N4CCOCC4)=N2)C=N1
Molecular Weight (g/mol) 377.42
Formula Weight 377.42
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.