missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ch-TOG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB500-182
This item is not returnable.
View return policy
Description
ch-TOG Polyclonal specifically detects ch-TOG in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| ch-TOG | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500 | |
| Q14008 | |
| CKAP5 | |
| A recombinant segment of the C-terminal domain of human ch-TOG [UniProt# Q14008] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Mammalia | |
| Antisera |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| Ch-TOG, CHTOG, Colonic and hepatic tumor over-expressed gene protein, cytoskeleton associated protein 5, cytoskeleton-associated protein 5, EC 2.7.7.6, EC 3.1.1.29, KIAA0097FLJ35359, MSPS, TOG, TOGp | |
| Rabbit | |
| 200 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
| 9793 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction