missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ch-TOG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92555-0.02ml
This item is not returnable.
View return policy
Description
ch-TOG Polyclonal antibody specifically detects ch-TOG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| ch-TOG | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:500 - 1:1000 | |
| Ch-TOG, CHTOG, Colonic and hepatic tumor over-expressed gene protein, cytoskeleton associated protein 5, cytoskeleton-associated protein 5, EC 2.7.7.6, EC 3.1.1.29, KIAA0097FLJ35359, MSPS, TOG, TOGp | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1763-2032 of human CKAP5 (NP_001008938.1). LLHTLCKLKGPKILDHLTMIDNKNESELEAHLCRMMKHSMDQTGSKSDKETEKGASRIDEKSSKAKVNDFLAEIFKKIGSKENTKEGLAELYEYKKKYSDADIEPFLKNSSQFFQSYVERGLRVIEMEREGKGRISTSTGISPQMEVTCVPTPTSTVSSIGNTNGEEVGPSVYLERLKILRQRCGLDNTKQDDRPPLTSLLSKPAVPTVASSTDMLHSKLSQLRESREQHQHSDLDSNQTHSSGTVTSSSSTANIDDLKKRLERIKSSRK | |
| 0.02 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Signal Transduction | |
| 9793 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction