missing translation for 'onlineSavingsMsg'
Learn More

CGS 9343B, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18015382
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015382 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18015382 Supplier Tocris Bioscience Supplier No. 2255/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calmodulin antagonist

Calmodulin antagonist; inhibits calmodulin-stimulated cAMP phosphodiesterase activity with an IC50 of 3.3 nM. Also prevents estrogen-induced transcription activation by ER, reversibly blocks voltage-activated Na+, Ca2+ and K+ currents in PC12 cells and inhibits nAChR.
TRUSTED_SUSTAINABILITY

Specifications

CAS 109826-27-9
Molecular Formula C30H32N4O6
MDL Number MFCD04113002
Quantity 10 mg
Synonym 1,3-dihydro-1-1-4-methyl-4h,6h-pyrrolo 1,2-a 4,1-benzoxazepin-4-yl methyl-4-piperidinyl-2h-benzimidazol-2-one maleate
InChI Key NGODOSILXOFQPH-BTJKTKAUNA-N
SMILES OC(=O)\C=C/C(O)=O.CC1(CN2CCC(CC2)N2C(=O)NC3=CC=CC=C23)OCC2=CC=CC=C2N2C=CC=C12
IUPAC Name (2Z)-but-2-enedioic acid; 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular Weight (g/mol) 544.61
PubChem CID 14171586
Formula Weight 544.61
Percent Purity >98%
Chemical Name or Material CGS 9343B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.