missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CGS 15943
Description
CGS 15943 is a potent adenosine receptor antagonist (Ki values are 3.5, 4.2, 16 and 51 nM for human A1, A2A, A2B and A3 receptors respectively). Inhibits the catalytic subunit of the class IB PI3K isoform p110γ (IC50 = 1.1 μM). Blocks HCC and HDAC cell proliferation in vitro via inhibition of the PI3K/Akt pathway; reduces proliferation of ER+ breast cancer cells. Also acts as a BMP-4 mimetic to stimulate osteogenic differentiation. Orally active in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 285.69 |
| Purity | >99% |
| Molecular Weight (g/mol) | 285.69 |
| Chemical Name or Material | CGS 15943 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?