missing translation for 'onlineSavingsMsg'
Learn More

CGS-12066A Maleate Salt, TRC

Product Code. 30362149
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362149 25 mg -
30350952 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362149 Supplier Toronto Research Chemicals Supplier No. TRCC29179325MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1350965-83-1
Molecular Formula C17H17F3N4 . C4H4O4
Molecular Weight (g/mol) 450.4
Synonym CGS-12066A Maleate Salt, 4-(4-Methyl-1-piperazinyl)-7-(trifluoromethyl)-pyrrolo[1,2-a]quinoxaline (2Z)-2-Butenedioate
IUPAC Name (Z)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
SMILES CN1CCN(CC1)c2nc3cc(ccc3n4cccc24)C(F)(F)F.OC(=O)\C=C/C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H17F3N4.C4H4O4/c1-22-7-9-23(10-8-22)16-15-3-2-6-24(15)14-5-4-12(17(18,19)20)11-13(14)21-16;5-3(6)1-2-4(7)8/h2-6,11H,7-10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 450.1515
Chemical Name or Material 4-(4-Methyl-1-piperazinyl)-7-(trifluoromethyl)-pyrrolo[1,2-a]quinoxaline (2Z)-2-Butenedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.