missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CGRRF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551779

Product Code. 15789730

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IYDIISMVSV IHIPDRTYKL SCRILYQYLL LAQGQFHDLK QLFMSANNNF TPSNNSSSEE KNTDRSLLEK VGLSESEVEP SEENSKDCVV CQNGTVNWVL LPCRHTCLCD GCVKYFQQCP MCRQFVQESF ALCSQKEQDK DKPK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

CGRRF1 is able to inhibit growth in several cell lines.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CGRRF1
Polyclonal
Unconjugated
CGRRF1
1110038G02Rik; 1810009H17Rik; cell growth regulator with ring finger domain 1; cell growth regulator with RING finger domain protein 1; Cell growth regulatory gene 19 protein; cell growth regulatory with ring finger domain; CGR19; Cgrrf1; RING finger protein 197; RNF197
Rabbit
Antigen affinity chromatography
RUO
10668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q99675
CGRRF1
Recombinant protein corresponding to Human CGRRF1. Recombinant protein control fragment (Product #RP-89080).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.