missing translation for 'onlineSavingsMsg'
Learn More

CGRP1 Antibody (4), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18370095 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370095 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370095 Supplier Novus Biologicals Supplier No. NBP300519JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CGRP1 Monoclonal antibody specifically detects CGRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGRP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I
Host Species Mouse
Immunogen Synthetic human alpha-CGRP adsorbed onto aluminum hydroxide gel. The epitope lies within residues 19-37 of human alpha-CGRP.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Signaling, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.