missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGRP Monoclonal Antibody (4901), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA547687
This item is not returnable.
View return policy
Description
Specific for Calcitonin Gene Related Peptide. Also recognizes rat beta-CGRP and human alpha-CGRP (10-fold less sensitivity compared to rat alpha-CGRP). Suggested positive controls are dorsal root ganglia, pancreas and gut.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CGRP | |
Monoclonal | |
Lot-Specific | |
10mM HEPES with 100μg/mL BSA, 150mM NaCl, 50% glycerol and no preservative; pH 7.5 | |
P01256, P01257 | |
Calca | |
Purified rat alpha-CGRP. | |
100 μL | |
Primary | |
Rat | |
Liquid |
Immunocytochemistry | |
4901 | |
Unconjugated | |
Calca | |
alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 | |
Mouse | |
Protein G | |
RUO | |
24241 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |