missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGRP Monoclonal Antibody (4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0260402
This item is not returnable.
View return policy
Description
ABS 026-04-02 detects CGRP alpha in human samples. ABS 026-04-02 has been successfully used in ELISA and immunohistochemistry procedures.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CGRP | |
| Monoclonal | |
| 1.03 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P01258, P06881 | |
| Calca | |
| Synthetic peptide corresponding to residues 19-37 of human alpha-CGRP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| 4 | |
| Unconjugated | |
| Calca | |
| alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 | |
| Mouse | |
| Protein A | |
| RUO | |
| 796 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction