missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGREF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99923-100ul
This item is not returnable.
View return policy
Description
CGREF1 Polyclonal antibody specifically detects CGREF1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CGREF1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin | |
| Produced in rabbits immunized with purified, recombinant Human CGREF1 (Uniprot#: Q99674-1; Met1-Ile301) | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 10669 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction