missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CGREF1 Polyclonal specifically detects CGREF1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | CGREF1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant CGREF1 (Q99674-1; Met1-Ile301). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?