missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGP71683 hydrochloride, MedChemExpress
Description
CGP71683 hydrochloride is a competitive neuropeptide Y5 receptor antagonist with a Ki of 1.3 nM, and shows no obvious activity at Y1 receptor (Ki, >4000 nM) and Y2 receptor (Ki, 200 nM) in cell membranes.
Specifications
Specifications
| Chemical Name or Material | CGP71683 hydrochloride |
| CAS | 192322-50-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H30ClN5O2S |
| Quantity | 50 mg |
| Synonym | CGP71683A |
| Solubility Information | DMSO : 160 mg/mL (312.46 mM; Need ultrasonic) |
| SMILES | O=S(C1=C2C=CC=CC2=CC=C1)(NC[C@H]3CC[C@H](CNC4=NC(N)=C5C=CC=CC5=N4)CC3)=O.Cl |
| Molecular Weight (g/mol) | 512.07 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?