missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CGP 78608 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective NMDA antagonist that acts through the glycine site (IC50 = 5 nM). Displays > 500-fold selectivity over kainate and AMPA receptors (IC50 values are 2.7 and 3 μM respectively). Also potentiates GluN1/GluN3A-mediated glycine currents (EC50 = 26.3 nM). Anticonvulsant in vivo following systemic administration.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 414.58 |
| Purity | >98% |
| Molecular Weight (g/mol) | 414.577 |
| Chemical Name or Material | CGP 78608 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction