missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGP 57380, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CGP 57380 is a cell-permeable pyrazolo-pyrimidine compound that acts as a selective inhibitor of Mnk1 with IC50 of 2.2 μM, but has no inhibitory activity against p38, JNK1, ERK1/2, PKC, or Src-like kinases.
Specifications
Specifications
| Chemical Name or Material | CGP 57380 |
| CAS | 522629-08-9 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C11H9FN6 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 6 mg/mL (24.57 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | NC1=NC=NC2=C1C(NC3=CC=C(F)C=C3)N=N2 |
| Molecular Weight (g/mol) | 244.23 |
| Formula Weight | 244.23 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction