missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CGP 53353
GREENER_CHOICE

Product Code. 18044092
Change view
Click to view available options
:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18044092 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18044092 Supplier Tocris Bioscience™ Supplier No. 2442/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of PKCβII

CGP 53353 is a selective inhibitor of PKCβII (IC50 values are 0.41 and 3.8 μM for PKCβII and PKCβI respectively). Also inhibits prionogenic Sup35 fibrillization (IC50 ∽ 3.4 μM) and inhibits de novo Aβ42 assembly in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type CGP 53353
Purity >94%
Molecular Weight (g/mol) 365.34
Inhibitors CGP 53353

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.