missing translation for 'onlineSavingsMsg'
Learn More

CGP 46381, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18034792
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18034792 10 mg 10mg
18065642 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034792 Supplier Tocris Bioscience Supplier No. 1247/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective GABAB antagonist; brain penetrant

Brain penetrant, selective GABAB receptor antagonist (IC50 = 4.9 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123691-14-5
Molecular Formula C10H22NO2P
Molecular Weight (g/mol) 219.265
InChI Key XOESDNIUAWGCLU-UHFFFAOYSA-N
Synonym 3-aminopropyl-cyclohexylmethylphosphinic acid, 3-aminopropyl cyclohexylmethyl phosphinic acid, 3-aminopropyl-cyclohexylmethyl-phosphinic acid, phosphinic acid, p-3-aminopropyl-p-cyclohexylmethyl, tocris-1247, biomol-nt_000238, d0b8se, 3-aminopropyl-cyclohexylmethyl phosphinic acid, phosphinic acid, 3-aminopropyl cyclohexylmethyl
PubChem CID 130022
IUPAC Name 3-aminopropyl(cyclohexylmethyl)phosphinic acid
SMILES C1CCC(CC1)CP(=O)(CCCN)O

Specifications

Quantity 10 mg
Formula Weight 219.26
Percent Purity >98%
Chemical Name or Material CGP 46381
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.