missing translation for 'onlineSavingsMsg'
Learn More

CGP-42112, MedChemExpress

Product Code. 16447867
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447867 5 mg 5mg
16441249 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447867 Supplier Medchem Express Supplier No. HY12405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CGP-42112 (CGP-42112A) is a potent Angiotensin-II subtype 2 receptor(AT2 R) agonist.

CGP-42112 (CGP-42112A) is a potent Angiotensin-II subtype 2 receptor(AT2 R) agonist.

Specifications

Chemical Name or Material CGP-42112
CAS 127060-75-7
Color White
Purity Grade Notes Research
Molecular Formula C52H69N13O11
Quantity 5 mg
Synonym CGP42112A
Solubility Information DMSO : 100 mg/mL (95.04 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H](C(O)=O)NC([C@H]1N(C([C@H](CC2=CNC=N2)NC([C@H](CCCCNC([C@H](CCCNC(N)=N)NC(OCC3=CC=CC=C3)=O)=O)NC([C@H](CC4=CC=C(C=C4)O)NC(C5=CC=CN=C5)=O)=O)=O)=O)CCC1)=O
Molecular Weight (g/mol) 1052.19
Formula Weight 1052.19
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.