missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CGP 37157

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective antagonist of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 324.22 |
| Purity | >99% |
| Molecular Weight (g/mol) | 324.219 |
| Chemical Name or Material | CGP 37157 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction