missing translation for 'onlineSavingsMsg'
Learn More

CGP 25454A, MedChemExpress

Product Code. 16415767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16415767 5 mg 5mg
16433028 10 mg 10mg
16409596 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16415767 Supplier Medchem Express Supplier No. HY100454
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CGP 25454A is a novel and selective presynaptic dopamine autoreceptor antagonist.

CGP 25454A is a novel and selective presynaptic dopamine autoreceptor antagonist.

Specifications

Chemical Name or Material CGP 25454A
CAS 104391-26-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H21Cl2N3O2
Quantity 5 mg
Solubility Information DMSO : ≥ 3.5 mg/mL (10.11 mM)
SMILES O=C(NCCN(CC)CC)C1=CC(C#N)=C(Cl)C=C1OC.[H]Cl
Molecular Weight (g/mol) 346.25
Formula Weight 346.25
Percent Purity 99.46%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.