missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGNL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584806
This item is not returnable.
View return policy
Description
Immunogen sequence: NRATATSPDS GAKKISVKTF PSASNTQATP DLLKGQQELT QQTNEETAKQ ILYNYLKEGS TDNDDATKRK VNLVFEKIQT LKSRAAGSAQ GNNQACNSTS EVKDLLEQKS KLT Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 85%.
Paracingulin (CGNL1, cingulin-like 1, FLJ14957, JACOP, KIAA1749) is novel protein with similar domain organization and significant sequence homology to cingulin, one of the tight junction (TJ) molecules. Paracingulin may play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
Specifications
| CGNL1 | |
| Polyclonal | |
| Unconjugated | |
| CGNL1 | |
| 4933421H10Rik; 9930020M10Rik; AI503810; CGNL1; cingulin like 1; cingulin-like 1; cingulin-like protein 1; JACOP; Junction-associated coiled-coil protein; Kiaa1749; Paracingulin; PCING | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84952 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0VF96 | |
| CGNL1 | |
| Recombinant protein corresponding to Human CGNL1. Recombinant protein control fragment (Product #RP-107682). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction