Learn More
Invitrogen™ cGKI Polyclonal Antibody
Description
Immunogen sequence: MGTLRDLQYA LQEKIEELRQ RDALIDELEL ELDQKDELIQ KLQNELDKYR SVIRPATQQA QKQSASTLQG EPRTKRQAIS AEPTAFDIQD LSHVTLPFYP KSPQSKDLIK EAILDNDFMK NLELSQIQEI VDCMYPVEYG KDSCIIKEGD VGSLVYVMED GKVEVTKEGV KLCTMGPGKV FGELAILYNC TRTATVKTLV NVKLWAIDRQ CFQTIMMRTG LIKHTEYMEF LKSVPTFQSL PEEILSKLAD VLEETHYENG EYIIRQGARG DTFFIISKGT VNVTREDSPS EDPVFLRTLG; Positive Samples: HepG2; Cellular Location: Cytoplasm.
Specifications
Specifications
| Antigen | cGKI |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.85 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRKG1 |
| Gene Accession No. | P0C605, Q13976 |
| Gene Alias | AAT8; AW125416; cGK; cGK 1; CGK 1 alpha; cGK 1 beta; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; cGMP dependent protein kinase type 1; cGMP kinase type I alpha; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase 1, alpha isozyme; cGMP-dependent protein kinase 1, beta isozyme; cGMP-dependent protein kinase I; Gm19690; OTTHUMP00000019618; PKG; Pkgi; Prkg1; PRKG1B; PRKGR1A; PRKGR1B; protein kinase cGMP-dependent 1; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type 1; protein kinase, cGMP-dependent, type I; RP11-346D6.1; Unknown (protein for MGC:134126); unnamed protein product |
| Gene Symbols | PRKG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.