missing translation for 'onlineSavingsMsg'
Learn More

cGK2/PRKG2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18426819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426819 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426819 Supplier Novus Biologicals Supplier No. NBP305940JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

cGK2/PRKG2 Polyclonal antibody specifically detects cGK2/PRKG2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Target Species Human
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.