missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGI-62 Monoclonal Antibody (OTI4B5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526080
This item is not returnable.
View return policy
Description
CGI-62 Monoclonal Antibody for Western Blot, IHC (P)
The CGI-62 gene encodes a hypothetical protein located on chromosome 8.
Specifications
| CGI-62 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96GY0 | |
| ZC2HC1A | |
| Full length human recombinant protein of FAM164A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B5 | |
| Unconjugated | |
| ZC2HC1A | |
| 3110050N22Rik; AI790358; AU023959; C14H8ORF70; C8orf70; CGI-62; Fam164a; family with sequence similarity 164, member A; protein FAM164A; RGD1311970; UPF0418 protein FAM164A; Zc2hc1a; Zinc finger C2HC domain-containing protein 1A; zinc finger C2HC-type containing 1A; zinc finger, C2HC-type containing 1A | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 51101 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction