missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI 62 Antibody (OTI4C1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71896AF594
This item is not returnable.
View return policy
Description
CGI 62 Monoclonal specifically detects CGI 62 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| CGI 62 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZC2HC1A | |
| Full length recombinant protein of human CGI 62 (NP_057094) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4C1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| C8orf70, CGI-62, chromosome 8 open reading frame 70, family with sequence similarity 164, member A, hypothetical protein LOC51101 | |
| Mouse | |
| 0.1 mL | |
| 51101 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction