missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Abnova™ CGI-38 293T Cell Transient Overexpression Lysate (Denatured)
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
Förpackningsstorlek
100µL
Tekniske data
Tekniske data
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | CGI-38 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_015964 |
| Molecular Weight (g/mol) | Theoretical M.W. 19kDa |
| Plasmid | pCMV-CGI-38 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-CGI-38 antibody (Mfr. No. H00051673-B02P) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 100UL CGI-38 293T Cell Transient Overexpression Ly
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion