missing translation for 'onlineSavingsMsg'
Learn More

CGI-1746, MedChemExpress

Product Code. p-200100232
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494898

Brand: Medchem Express HY11999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CGI-1746 is a potent and highly selective inhibitor of the Btk with IC50 of 1.9 nM.

CGI-1746 is a potent and highly selective inhibitor of the Btk with IC50 of 1.9 nM.

Specifications

Chemical Name or Material CGI-1746
Physical Form Solid
CAS 910232-84-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C34H37N5O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (86.25 mM)
SMILES O=C(C1=CC=C(C=C1)C(C)(C)C)NC2=CC=CC(C(N=C3NC4=CC=C(C=C4)C(N5CCOCC5)=O)=CN(C3=O)C)=C2C
Molecular Weight (g/mol) 579.69
Formula Weight 579.69
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.