missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI-128 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92518-0.02ml
This item is not returnable.
View return policy
Description
CGI-128 Polyclonal antibody specifically detects CGI-128 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CGI-128 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CGI-128, family with sequence similarity 96, member B, MIP18, mitotic spindle-associated MMXD complex subunit MIP18, MSS19-interacting protein of 18 kDa, Protein FAM96B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human FAM96B (NP_057146.1). MVGGGGVGGGLLENANPLIYQRSGERPVTAGEEDEQVPDSIDAREIFDLIRSINDPEHPLTLEELNVVEQVRVQVSDPESTVAVAFTPTIPHCSMATLIGLSIKVKLLRSLPQRFKMDVHITPGTHASEHAVNKQLADKERVAAALENTHLLEVVNQCLSARS | |
| 0.02 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 51647 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction