missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CGGBP1 influences expression of the FMR1 gene which is associated with the fragile X mental retardation syndrome by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.
Specifications
Specifications
| Antigen | CGGBP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CGGBP1 |
| Gene Accession No. | Q8BHG9, Q9UFW8 |
| Gene Alias | 20 kDa CGG binding protein, CGG binding protein 1, CGGBP, CGGBP1, p20 CGGBP, p20 CGGBP DNA binding protein |
| Gene Symbols | Cggbp1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?