missing translation for 'onlineSavingsMsg'
Learn More

CGGBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16897273 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16897273 20 μL 20µL
16887273 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16897273 Supplier Proteintech Supplier No. 107161AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CGGBP1 influences expression of the FMR1 gene which is associated with the fragile X mental retardation syndrome by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.

Specifications

Antigen CGGBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CGGBP1
Gene Accession No. Q8BHG9, Q9UFW8
Gene Alias 20 kDa CGG binding protein, CGG binding protein 1, CGGBP, CGGBP1, p20 CGGBP, p20 CGGBP DNA binding protein
Gene Symbols Cggbp1
Host Species Rabbit
Immunogen CGGBP1 Fusion Protein Ag1129
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106143, 288353, 8545
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.