missing translation for 'onlineSavingsMsg'
Learn More

CGBP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA589228

Product Code. 16507201

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MEGDGSDPEP PDAGEDSKSE NGENAPIYCI CRKPDINCFM IGCDNCNEWF HGDCIRITEK MAKAIREWYC RECREKDPKL EIRYRHKKSR ERDGNERDSS EPRDEGGGRK RPVPDPDLQR RAGSGTGVGA MLARGSASPH KSSPQPLVAT PSQHHQQQQQ QIKRSARMCG ECEACRRTED CGHCDFCRDM KKFGGPNKIR QKCRLRQCQL; Positive Samples: HepG2; Cellular Location: Nucleus speckle

Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CGBP
Polyclonal
Unconjugated
CXXC1
2410002I16Rik, 5830420C16Rik, AI426635, CFP 1, CFP1, CGBP, CpG binding protein, cpG-binding protein, cpG-binding protein, CXXC-type zinc finger protein 1, CXXC 1, CXXC finger 1, CXXC finger 1 (PHD domain), CXXC finger protein 1, Cxxc1, CXXC-type zinc finger protein 1, DNA-binding protein with PHD finger and CXXC domain, hCGBP, HsT2645, PCCX 1, Pccx1, PHD finger and CXXC domain-containing protein 1, PHF 18, PHF18, SPP 1, SPP1, ZCGPC 1, ZCGPC1, zinc finger, CpG binding-type containing 1
Rabbit
Affinity Chromatography
RUO
291440, 30827, 74322
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Q9CWW7, Q9P0U4
CXXC1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human CXXC1 (NP_0010951241)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.