missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130202
This item is not returnable.
View return policy
Description
Expected to cross react with mouse samples due to sequence homology. A suggested positive control for this product is human spleen cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
Specifications
| CGBP | |
| Polyclonal | |
| Unconjugated | |
| CXXC1 | |
| 2410002I16Rik; 5830420C16Rik; AI426635; CFP 1; CFP1; CGBP; CpG binding protein; cpG-binding protein; cpG-binding protein; CXXC-type zinc finger protein 1; CXXC 1; CXXC finger 1; CXXC finger 1 (PHD domain); CXXC finger protein 1; Cxxc1; CXXC-type zinc finger protein 1; DNA-binding protein with PHD finger and CXXC domain; hCGBP; HsT2645; PCCX 1; Pccx1; PHD finger and CXXC domain-containing protein 1; PHF 18; PHF18; SPP 1; SPP1; ZCGPC 1; ZCGPC1; zinc finger, CpG binding-type containing 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 30827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q9P0U4 | |
| CXXC1 | |
| Synthetic peptides, CREKDPKLEIRYRHKKSR and IQHDPLTTDLRSSADR corresponding to amino acids 73-90 and 641-656 of Human CGBP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction