missing translation for 'onlineSavingsMsg'
Learn More

CGB5 Antibody, FITC, Novus Biologicals™

Product Code. 18363186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363186 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363186 Supplier Novus Biologicals Supplier No. NBP300335F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CGB5 Polyclonal specifically detects CGB5 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGB5
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CGB3, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 5 subunit, chorionic gonadotropin, beta polypeptide 5, HCG, MGC119822
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CGB5 (P01233-1; Met1-Gln165).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 93659
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.