Learn More
cGAS Rabbit, anti-Human, Clone: 1H1, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82869-1-RR-100UL
This item is not returnable.
View return policy
Description
The cGAS antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human cGAS. This antibody recognizes human antigen. The cGAS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| cGAS | |
| 1H1 | |
| C6orf150, cGAS, h-cGAS, MB21D1 | |
| 100 μL | |
| 115004 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |