Learn More
cGAS Mouse, anti-Human, Clone: 2G2C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68640-1-IG-20UL
This item is not returnable.
View return policy
Description
The cGAS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human cGAS. This antibody recognizes human antigen. The cGAS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| cGAS | |
| 2G2C5 | |
| C6orf150, cGAS, h-cGAS, MB21D1 | |
| 20 μL | |
| 115004 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |