missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cGAMP, MedChemExpress
Description
cGAMP (Cyclic GMP-AMPP) functions as an endogenous second messenger in metazoans and triggers interferon production in response to cytosolic DNA. cGAMP activates stimulator of interferon genes (STING), which activates a signaling cascade leading to the production of type I interferons and other immune mediators.
Specifications
Specifications
| Chemical Name or Material | cGAMP |
| CAS | 849214-04-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H24N10O13P2 |
| Quantity | 1 mg |
| Synonym | Cyclic GMP-AMP 3',3'-cGAMP |
| Solubility Information | H2O : 20 mg/mL (29.66 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble) |
| SMILES | O=P(O[C@H]1[C@@H](O)[C@H](N2C=NC3=C2N=CN=C3N)O[C@@H]1COP4(O)=O)(O)OC[C@@H]5[C@@H](O4)[C@@H](O)[C@H](N6C=NC7=C6N=C(N)NC7=O)O5 |
| Molecular Weight (g/mol) | 674.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?