missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CFTRinh 172

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Voltage-independent, selective CFTR chloride channel blocker (Ki = 300 nM) that alters channel gating. Blocks intestinal fluid secretion induced by cholera toxin and Escherichia coli and suppresses cyst growth in animal models of polycystic kidney disease. Orally active. Inhibits mitochondrial respiration and increases reactive oxygen species (ROS) production independently of CFTR in several cell lines.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 409.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 409.397 |
| Chemical Name or Material | CFTRinh 172 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction